Antimony powder, -200 mesh, 99.999% (metal basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1464003

Quantity 1g

 View more versions of this product

Catalog No. AA1464003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-36-0
    Physical Form Powder
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00134030
    Chemical Name or Material Antimony

    Specifications

    Quantity 1g
    Color Gray
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 231-146-5
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.999% (Metals basis)
    Appearance Gray powder
    MDL Number MFCD00134030
    CAS 7440-36-0
    Health Hazard 1 H301-H331-H411-H401: Toxic if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
    Chemical Name or Material Antimony

    H301-H331-H411-H401: Toxic if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit