Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony rod, 12.7mm (0.50in) dia x 10cm (3.9in) long, 99.8% (metals basis)

Catalog No. p-7048296
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10098KG 1 Pc.
AA10098KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10098KG Supplier Thermo Scientific Chemicals Supplier No. 010098KG
Only null left
Add to Cart

CAS: 7440-36-0 | Sb | 121.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]

Specifications

Quantity 1 Pc.
Odor Odorless
Weight ≈81.2g (0.178lbs)/10cm piece
Form Cast Rod
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Antimony rod, 12.7mm (0.50 in.) dia. x 10cm (3.9 in.) long
EINECSNumber 231-146-5
RTECSNumber CC4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.