Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony Trioxide (Powder/Certified), Fisher Chemical

Catalog No. A860500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A860-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A860-500

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 1550°C
Melting Point 656°C
Color White
CAS 1309-64-4
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula O3Sb2
MDL Number MFCD00011214
Synonym Antimonous Oxide
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]
IUPAC Name diantimony(3+) trioxidandiide
Molecular Weight (g/mol) 291.52
Formula Weight 291.52
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Antimony Trioxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.