Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(V) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific™

Catalog No. AA40128A4
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40128A4 2.5 kg
AA4012836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40128A4 Supplier Thermo Scientific Chemicals Supplier No. 40128A4

missing translation for 'validMainframeMsg'

Flame retardant

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-60-9
Molecular Formula O5Sb2
Molecular Weight (g/mol) 323.515
MDL Number MFCD00011216
InChI Key LJCFOYOSGPHIOO-UHFFFAOYSA-N
Synonym antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40
PubChem CID 14813
IUPAC Name (dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
SMILES O=[Sb](=O)O[Sb](=O)=O

Specifications

Physical Form Particles in Liquid
Quantity 2.5 kg
Formula Weight 323.5
Concentration or Composition (by Analyte or Components) 30% in H2O
Odor Odorless
Chemical Name or Material Antimony(V) oxide, Colloidal dispersion
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 215-237-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.