Learn More
Fluidigm Corp Anti-Mouse CD180/RP105 (RP/14)-162Dy 25 Tests
Supplier: Fluidigm Corp 3162031C
CD180, also known as RP105 or Ly64, belongs to the TLR family and is expressed on most B cells, monocytes/macrophages, and dendritic cells.