Learn More
Fluidigm Corp Anti-Mouse CD28 (37.51)-151Eu 25 Tests
SDP
Supplier:  Fluidigm Corp 3151005C
Encompass
CD28 binds to CD80 and CD86, can be expressed on T cells, NK cell subsets, and plasma cells, and provides co-stimulatory signals.
Catalog No. 50-196-2738