Learn More
American Research Products Inc AntiMouse CD28 In Vivo Ab
Catalog No. 5000306674
Encompass
Anti-Mouse CD28 In Vivo Antibody - Low Endotoxin is a in vivo antibody with mouse reactivity produced in syrian hamster Available in 100 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles
Catalog No. 50-003-06674 Supplier American Research Products Inc Supplier No. IVMB005625MG
Anti-Mouse CD28 In Vivo Antibody - Low Endotoxin is a in vivo antibody with mouse reactivity produced in syrian hamster Available in 100 mg size with 7 - 14 business days lead time Ships via gel pack Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month For longer term storage aseptically aliquot in working volumes without diluting and store at -80 C Avoid Repeated Freeze Thaw Cycles