Learn More
Fluidigm Corp Anti-Mouse CD38 (90)-171Yb 25 Tests
SDP
Supplier:  Fluidigm Corp 3171007C
Encompass
CD38 is expressed at variable levels across most hematopoietic cells in the mouse, and it is down-regulated on mouse germinal center B cells and plasma cells. This is not the case for humans.
Catalog No. 50-196-3301