Learn More
Fluidigm Corp Anti-Mouse CD39 (24DMS1)-142Nd 100 Tests
SDP
Supplier:  Fluidigm Corp 3142005B
Encompass
CD39 is expressed in B cells, subsets of dendritic cells, most monocytes, and CD4+CD25+ immunosuppressive regulatory T (Treg) cells.
Catalog No. 50-196-2500