Learn More
Fluidigm Corp Anti-Mouse CD41 (MWReg30)-143Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3143009C
Encompass
CD41, also known as integrin ?IIb, is a transmembrane glycoprotein and is commonly used as a marker of platelets and megakaryocytes.
Catalog No. 50-196-2523