Learn More
Fluidigm Corp Anti-Mouse CD43 (S11)-146Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3146009C
Encompass
CD43, also known as leukosialin, is a highly glycosylated 125 kDa type I transmembrane protein with a mucin-like structure expressed in all immune cells, except naïve B lymphocytes.
Catalog No. 50-196-2605