Learn More
Fluidigm Corp Anti-Mouse CD5 (53-7.3)-146Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3146012C
Encompass
CD5, also known as Lyt-1, Ly-1, Tp67, T1, or Ly12, is a member of the scavenger receptor cysteine-rich (SRCR) protein superfamily. It is primarily expressed on thymocytes, peripheral T cells, and B-1 cells.
Catalog No. 50-196-2610