Learn More
Fluidigm Corp Anti-Mouse IgM (RMM-1)-151Eu 25 Tests
SDP
Supplier:  Fluidigm Corp 3151006C
Encompass
IgM is an immunoglobulin heavy chain isotype and is expressed intracellularly during early stages of B cell development and on the surface of mature B cells in the bone marrow and periphery.
Catalog No. 50-196-2740