Learn More
Fluidigm Corp Anti-Mouse MHC Class I (28-14-8)-144Nd 25 Tests
SDP
Supplier:  Fluidigm Corp 3144016C
Encompass
The 28-14-8 monoclonal antibody reacts with the mouse MHC class I, H-2Db, and cross-reacts with H-2Ld, H-2Dq, and/or H-2Lq. Binding of 28-14-8 to the alpha 3 domain of H-2Ld is not dependent on ?2-microglobulin.
Catalog No. 50-196-2564