Learn More
Fluidigm Corp Anti-Mouse/Rat CD40 (HM40-3)-161Dy 100 Tests
SDP
Supplier:  Fluidigm Corp 3161020B
Encompass
CD40, a member of the TNFR superfamily, is primarily expressed on B cells, macrophages, follicular dendritic cells, endothelial cells, fibroblasts, and at low levels on plasma cells.
Catalog No. 50-196-3022