Learn More
Leinco Technologies Inc Anti-Mouse Ly-6C (Lymphocyte antigen 6 complex, locus C) In Vivo GOLD; Clone HK1.4; Isotype: Rat IgG2c
SDP
Supplier:  Leinco Technologies Inc L136100 MG
Encompass_Preferred
Specificity: Clone HK1.4 recognizes an epitope on mouse Ly-6C. Antigen Distribution: Ly-6C is a marker of memory CD8+ T cells and is mainly expressed on bone marrow myeloid populations, monocytes/macrophages, neutrophils, endothelial cells, and some T cell subsets. Background: Ly-6C is a 14-17 kD GPI linked protein and a member of the Ly-6 family. Ly6 family members share structure homology throughout a characteristic cystein rich protein domain that incorporates O-linked carbohydrates. Development stage and activation play a role in Ly-6 expression that occurs on most hematopoietic cells. Applications: (IP, WB) Clone HK1.4 does not block the binding of clone RB6-8C5. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6504