Learn More
Enzo Life Sciences Antimycin A, (10mg), CAS Number: 1397-94-0
SDP
Catalog No. NC9061331
Encompass_Preferred
Antibiotic. Induces apoptosis, which is not prevented by the presence of Bcl-2. Inhibits mitochondrial electron transport specifically between cytochromes b and c1. Alternative Name: Antipiricullin, Virosin. CAS Number: 1397-94-0. MI: 14: 714 (A1) & 715 (A3). Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol (50mg/ml). Long Term Storage: -20°C
Catalog No. NC9061331 Supplier Enzo Life Sciences Supplier No. ALX380075M010
Antibiotic. Induces apoptosis, which is not prevented by the presence of Bcl-2. Inhibits mitochondrial electron transport specifically between cytochromes b and c1. Alternative Name: Antipiricullin, Virosin. CAS Number: 1397-94-0. MI: 14: 714 (A1) & 715 (A3). Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol (50mg/ml). Long Term Storage: -20°C