Learn More
Lkt Laboratories CAS 1397-94-0, Cytochrome C binding agent that inhibits electron transport chain activity, oxidative phosphorylation, and ATP synthesis.
SDP
Supplier:  Lkt Laboratories A5378
Encompass_Preferred
Antimycin A was initially produced by Streptomyces. Antimycin A binds cytochrome c reductase, inhibiting electron transport chain activity, oxidative phosphorylation, and ATP synthesis. Antimycin A is used in research models to study mitochondrial respiration and superoxide production.
Catalog No. NC2530043