Learn More
Medchemexpress LLC Antineoplaston A10 | 91531-30-5 | 99.8% | 100 MG
SDP
Catalog No. 5000338269
Encompass_Preferred
Antineoplaston A10 is an antineoplaston that inhibits the growth of human hepatoma cells by inducing apoptosis. It is for research use only and can be used in the study of liver cancer and breast cancer.
  • Inhibits the growth of human hepatoma cell lines HepG2 and HLE
  • Induces apoptosis
  • Targets Ras
  • Reduces bcl-2 expression in cytoplasm
  • Increases nuclear p53 expression
Catalog No. 50-003-38269 Supplier Medchemexpress LLC Supplier No. HY128553100MG
Antineoplaston A10 is an antineoplaston that inhibits the growth of human hepatoma cells by inducing apoptosis. It is for research use only and can be used in the study of liver cancer and breast cancer.
  • Inhibits the growth of human hepatoma cell lines HepG2 and HLE
  • Induces apoptosis
  • Targets Ras
  • Reduces bcl-2 expression in cytoplasm
  • Increases nuclear p53 expression