Learn More
Medchemexpress LLC Antioxidant 1010 | 6683-19-8 | 98.04% | 1177.66 | 50 G
SDP
Catalog No. 5000452344
Encompass_Preferred
Antioxidant 1010 exhibits antioxidant efficacy in plastic, rubber, and synthetic fiber. It is for research use only.
  • Exhibits antioxidant efficacy in plastic, rubber and synthetic fiber.
Catalog No. 50-004-52344 Supplier Medchemexpress LLC Supplier No. HYW01065350G
Antioxidant 1010 exhibits antioxidant efficacy in plastic, rubber, and synthetic fiber. It is for research use only.
  • Exhibits antioxidant efficacy in plastic, rubber and synthetic fiber.