Learn More
Medchemexpress LLC Antioxidant agent-1 | 136068-43-4 | 98.1% | C15H12O3 | 25 MG
SDP
Catalog No. 5000355988
Encompass_Preferred
Antioxidant agent-1 is a novel chalcone derivative, recognized as a potential antioxidant. This compound is suitable for laboratory research and substance manufacturing applications.
  • Functions as a potential antioxidant agent
  • Demonstrates inhibitory activity against 5-lipoxygenase (IC50 of 0.043 μM)
  • Exhibits inhibitory activity against cyclooxygenase (IC50 of 0.043 μM)
  • Effective in KB cells with a CC50 of 3.1 μM
  • Effective in MT4 cells with a CC50 of 1.7 μM
Catalog No. 50-003-55988 Supplier Medchemexpress LLC Supplier No. 25MGHY145104
Antioxidant agent-1 is a novel chalcone derivative, recognized as a potential antioxidant. This compound is suitable for laboratory research and substance manufacturing applications.
  • Functions as a potential antioxidant agent
  • Demonstrates inhibitory activity against 5-lipoxygenase (IC50 of 0.043 μM)
  • Exhibits inhibitory activity against cyclooxygenase (IC50 of 0.043 μM)
  • Effective in KB cells with a CC50 of 3.1 μM
  • Effective in MT4 cells with a CC50 of 1.7 μM