Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antioxidant agent-4 | 50901-34-3 | 96.0% | 168.15 | C8H8O4 | 25 MG
SDP

Catalog No. 5000209724
Encompass_Preferred

Antioxidant agent-4 is a small-molecule antioxidant (Compound 2) with reported antilipid peroxidative activity, supplied as a solid for research use. It is provided in multiple pack sizes and solution formats, with noted solubility in DMSO and storage recommendations for solid and solution forms.

  • Small-molecule antioxidant with antilipid peroxidative activity.
  • High purity (96.0%) suitable for research applications.
  • Chemical formula C8H8O4; molecular weight 168.15 g/mol.
  • Soluble in DMSO (100 mg/mL); in vivo formulation guidance available for several vehicles.
  • Available as solid in multiple pack sizes, including 25 MG.
  • Store solid at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09724 Supplier Medchemexpress LLC Supplier No. 25MGHYW068420
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent-4 is a small-molecule antioxidant (Compound 2) with reported antilipid peroxidative activity, supplied as a solid for research use. It is provided in multiple pack sizes and solution formats, with noted solubility in DMSO and storage recommendations for solid and solution forms.

  • Small-molecule antioxidant with antilipid peroxidative activity.
  • High purity (96.0%) suitable for research applications.
  • Chemical formula C8H8O4; molecular weight 168.15 g/mol.
  • Soluble in DMSO (100 mg/mL); in vivo formulation guidance available for several vehicles.
  • Available as solid in multiple pack sizes, including 25 MG.
  • Store solid at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.