Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antioxidant agent-4 | 50901-34-3 | 96.0% | 168.15 g/mol | C8H8O4 | 100mg
SDP

Catalog No. 5000210405
Encompass_Preferred

Antioxidant agent-4 is a small-molecule antioxidant reported to have antilipid peroxidative activity. It is supplied as a solid for laboratory research use only and is not intended for clinical or human use.

  • Small-molecule antioxidant with antilipid peroxidative activity.
  • Available in multiple pack sizes, including 100 mg.
  • Purity listed as 96.0%.
  • Molecular formula C8H8O4 and molecular weight 168.15 g/mol.
  • Intended for biochemical and cellular research applications.

Catalog No. 50-002-10405 Supplier Medchemexpress LLC Supplier No. HYW068420100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent-4 is a small-molecule antioxidant reported to have antilipid peroxidative activity. It is supplied as a solid for laboratory research use only and is not intended for clinical or human use.

  • Small-molecule antioxidant with antilipid peroxidative activity.
  • Available in multiple pack sizes, including 100 mg.
  • Purity listed as 96.0%.
  • Molecular formula C8H8O4 and molecular weight 168.15 g/mol.
  • Intended for biochemical and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.