Learn More
Enzo Life Sciences Antipain (25mg). CAS: 37682-72-7
SDP
Supplier:  Enzo Life Sciences ALX260093M025
Encompass_Preferred
Papain inhibitor. Inhibits papain and trypsin and to a small extent plasmin. Antipain is more specific for papain and trypsin than is leupeptin (ALX-260-009). The inhibitory potency of antipain is 100-fold higher than that of elastatinal. Acts by forming a hemiacetal adduct between its aldehyde group and the active serine of the proteinase. Alternative name: [(S)-1-Carboxy-2-phenylethyl]-carbamoyl-Arg-Val-arginal . 2HCl. Formula: C27H44N10O6 . 2HCl. MW: 604.7 . 72.9. Purity: ≥97% (HPLC). Solubility: Soluble in water, methanol, 100 % ethanol or acetonitrile. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8315