Learn More
Research Products International Corp Antipain Dihydrochloride, 5 Milligrams
SDP
Supplier:  Research Products International Corp A205600.005
Encompass_Preferred
Reversible cysteine and serine protease inhibitor of cathepsins A and B, papain and trypsin.
Working concentration: 1-100uM.
Catalog No. 50-488-492