Learn More
Carbosynth LLC. Antipain (Synthetic), 25mg, MW: 604.70 • 72.92, Molecular formula: C27H44N10O6 • 2HCl, CAS# 37682-72-7
SDP
Supplier:  Carbosynth LLC. IAP3996PI25MG
Encompass_Preferred
Synonyms: (S)-1-Carboxy-2-phenylethyl]-carbamoyl-Arg-Val-Arg-H • 2 HCl, Antipain • 2 HCl (Synthetic), [(S)-1-Carboxy-2-Phenylethyl]-Carbamoyl-L-Arginyl-L-Valyl-Argininal • dihydrochloride, (S)-1-Carboxy-2-phenylethyl]-carbamoyl-RVR-H, Application: Synthetic Product Inhibitor for Trypsin, u-PA, Papain, and Cathepsin A/B, Salt Form:, HCl salt, Storage Temperature: -20°C
Catalog No. 50-168-5812