Learn More
Fluidigm Corp Anti-Pan-Cytokeratin (AE1/AE3)-148Nd 25 µg
SDP
Supplier:  Fluidigm Corp 3148022D
Encompass
AE-1/AE-3 is a broad spectrum anti-pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.
Catalog No. 50-196-3465