Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antipyrine-d3 | 65566-62-3 | MFCD01074188 | 99.9% | 191.24 g/mol | C11H9D3N2O | 10 MG
SDP

Catalog No. 5000298863
Encompass_Preferred

Antipyrine-d3 is the deuterium-labeled internal standard of antipyrine supplied as a powder for analytical and pharmacokinetic studies. It is designed for isotope-dilution mass spectrometry and ADME/metabolic probe assays, providing a reliable tracer for quantitative LC-MS workflows.

  • Deuterium-labeled internal standard for antipyrine.
  • Supplied as a powder suitable for analytical use.
  • High purity (~99.9%) appropriate for LC-MS quantification.
  • Stable when stored as a powder: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Molecular formula C11H9D3N2O; molecular weight 191.24 g/mol.
  • Common applications: isotope-dilution assays, pharmacokinetic studies, and metabolic profiling.

Catalog No. 50-002-98863 Supplier Medchemexpress LLC Supplier No. HYB0171S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antipyrine-d3 is the deuterium-labeled internal standard of antipyrine supplied as a powder for analytical and pharmacokinetic studies. It is designed for isotope-dilution mass spectrometry and ADME/metabolic probe assays, providing a reliable tracer for quantitative LC-MS workflows.

  • Deuterium-labeled internal standard for antipyrine.
  • Supplied as a powder suitable for analytical use.
  • High purity (~99.9%) appropriate for LC-MS quantification.
  • Stable when stored as a powder: -20°C (up to 3 years) or 4°C (up to 2 years).
  • Molecular formula C11H9D3N2O; molecular weight 191.24 g/mol.
  • Common applications: isotope-dilution assays, pharmacokinetic studies, and metabolic profiling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.