Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, MilliporeSigma™ Supelco™

Catalog No. 111014868 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4868 25 g
11-101-4867 500 g
11-101-4869 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4868 Supplier MilliporeSigma Supelco Supplier No. A588225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antipyrine
Packaging Poly Bottle
CAS 60-80-0
Physical Form Neat
Linear Formula C11H12N2 O
Technique Liquid Chromatography
MDL Number MFCD00003146
Melting Point 109°C to 111°C
Synonym 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one; Phenazone
Type Analytical Standard
Industry Type Pharmaceutical
Molecular Weight (g/mol) 188.23
Quantity 25 g
Formula Weight 188.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.