Learn More
Med Vet International Zoetis Antirobe Aqua Drops, 25mg/mL, 20mL
Catalog No. 502828610
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Antirobe Aquadrops Liquid (clindamycin hydrochloride 25 mg/mL) is a palatable formulation intended for oral administration in dogs and cats. Indicated for the treatment of infections caused by susceptible strains of the designated microorganisms in the following conditions: DOGS Skin infections due to coagulase positive staphylococci. Deep wounds and abscesses due to Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens Dental infections due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens Osteomyelitis due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens CATS Skin infections due to Staphylococcus aureus, Staphylococcus intermedius and Streptococcus spp.
Catalog No. 50-282-8610 Supplier Med Vet International Supplier No. RXANTIROBE
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Antirobe Aquadrops Liquid (clindamycin hydrochloride 25 mg/mL) is a palatable formulation intended for oral administration in dogs and cats. Indicated for the treatment of infections caused by susceptible strains of the designated microorganisms in the following conditions: DOGS Skin infections due to coagulase positive staphylococci. Deep wounds and abscesses due to Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens Dental infections due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens Osteomyelitis due to Staphylococcus aureus, Bacteroides fragilis, Prevotella melaninogenicus, Fusobacterium necrophorum and Clostridium perfringens CATS Skin infections due to Staphylococcus aureus, Staphylococcus intermedius and Streptococcus spp.