Learn More
Leica Biosystems Anti-Roll Plate Assembly for Leica Cryostats, 70 mm, 50 μm
Catalog No. NC1921594
Encompass
Anti-roll plate assembly designed for use in cryostat anti-roll systems. Listed as 70 mm with a 50 μm specification, this plate fits into an interchangeable anti-roll frame and is intended for use in tissue sectioning applications in clinical histopathology, including Leica CM1860 cryostats.
  • Fits 70 mm anti-roll frame.
  • 50 μm specification for anti-roll system.
  • Replacement component for cryostat anti-roll systems.
  • Installs into an interchangeable frame for quick replacement.
  • Helps reduce tissue curling during cryostat sectioning.
Catalog No. NC1921594 Supplier Leica Biosystems Supplier No. 14041937258
Anti-roll plate assembly designed for use in cryostat anti-roll systems. Listed as 70 mm with a 50 μm specification, this plate fits into an interchangeable anti-roll frame and is intended for use in tissue sectioning applications in clinical histopathology, including Leica CM1860 cryostats.
  • Fits 70 mm anti-roll frame.
  • 50 μm specification for anti-roll system.
  • Replacement component for cryostat anti-roll systems.
  • Installs into an interchangeable frame for quick replacement.
  • Helps reduce tissue curling during cryostat sectioning.