Learn More
Med Vet International Hand Sanitizer 3M Avagard D 3oz Ethyl Alcohol Gel Bottle, 9221, Each
Catalog No. 502829021
Encompass_Preferred
3M™ Avagard™ D hand sanitizer contains 61% w/w ethyl alcohol to handle 99.99% of even antibiotic-resistant bacteria Formulated with moisturizers, it prevents skin breakdown and leaves hands soft after repeated use 3M™ Avagard™ D instant hand antiseptic with moisturizers dries quickly and is clean with no sticky residue Its compatibility with CHG products doesn't reduce its effectiveness It meets WHO, CDC and AORN efficacy guidelines
Catalog No. 50-282-9021 Supplier Med Vet International Supplier No. 406350EA
3M™ Avagard™ D hand sanitizer contains 61% w/w ethyl alcohol to handle 99.99% of even antibiotic-resistant bacteria Formulated with moisturizers, it prevents skin breakdown and leaves hands soft after repeated use 3M™ Avagard™ D instant hand antiseptic with moisturizers dries quickly and is clean with no sticky residue Its compatibility with CHG products doesn't reduce its effectiveness It meets WHO, CDC and AORN efficacy guidelines