Learn More
Med Vet International Hand Sanitizer 3M Avagard 1,000 mL Ethyl Alcohol Foaming Pump Bottle, 9322A, 5/Case
Catalog No. 502831265
Encompass_Preferred
1000 mL pump bottle Kills greater than 99.999% of harmful bacteria in 15 seconds (in vitro) Helps maintain skin condition Latex glove and CHG compatible USDA Certified Biobased product Fragrance-free
Catalog No. 50-283-1265 Supplier Med Vet International Supplier No. 800380CS
1000 mL pump bottle Kills greater than 99.999% of harmful bacteria in 15 seconds (in vitro) Helps maintain skin condition Latex glove and CHG compatible USDA Certified Biobased product Fragrance-free