Learn More
Med Vet International Hand Sanitizer 3M Avagard D 16oz Ethyl Alcohol Gel Pump Bottle, 9222, Each
Catalog No. 502829180
Encompass_Preferred
3M™ Avagard™ hand antiseptic kills 99.999% of pathogens for effective protection It meets the guidelines of WHO, AORN and CDC, which highlights its efficacy It has moisturizers to ensure the skin doesn't dry out, making it ideal for regular use The sanitizer is fast drying and residue-free for convenient comfort The 16-ounce bottle has a hand pump for dispensing
Catalog No. 50-282-9180 Supplier Med Vet International Supplier No. 406351EA
3M™ Avagard™ hand antiseptic kills 99.999% of pathogens for effective protection It meets the guidelines of WHO, AORN and CDC, which highlights its efficacy It has moisturizers to ensure the skin doesn't dry out, making it ideal for regular use The sanitizer is fast drying and residue-free for convenient comfort The 16-ounce bottle has a hand pump for dispensing