Learn More
Med Vet International Antiskid Drill Bit, 5/32”, 4.5mm, Length 13cm, J0372GX, Each
Catalog No. 5000581728
Encompass_Preferred
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.
Catalog No. 50-005-81728 Supplier Med Vet International Supplier No. JOR372GX
Add to Cart
Add to Cart
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.