Learn More
Med Vet International Antiskid Drill Bit, 5/64”, 2.0mm x 10cm, J0372BX, Each
Catalog No. 5000582047
Encompass_Preferred
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.
Catalog No. 50-005-82047 Supplier Med Vet International Supplier No. JOR372BX
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.