Learn More
Med Vet International Antiskid Drill Bit, J0372CX, 3/32in, 2.5mm x 11.5cm, Each
Catalog No. 5000581722
Encompass_Preferred
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.
Catalog No. 50-005-81722 Supplier Med Vet International Supplier No. JOR372CX
These drill bits have a sharp needle point tip that penetrates the bone for easier drilling. It helps avoid skipping over the bone especially if drilling at an angle other than 90°. Made from hard 420 stainless steel.