Learn More
ACL Inc ACL Plastic & Glass Cleaner, 14oz, 12/case
Catalog No. NC1610383
Encompass_Preferred
This foam cleaner is ideal to use on computer monitors and housings. It keeps observation windows on production equipment in pristine condition longer than other cleaners and does not contain CFC's or HCFC's.Controls static while cleaning and polishing. Effectively removes handling oils, finger soils, and nicotine stains. Prevents attraction of dust and dirt. Compatible with plastics, glass, and stainless steel. Foaming spray won't run or drip; ideal for optical surfaces. RoHS compliant.
Catalog No. NC1610383 Supplier ACL Inc Supplier No. 8670
This foam cleaner is ideal to use on computer monitors and housings. It keeps observation windows on production equipment in pristine condition longer than other cleaners and does not contain CFC's or HCFC's.Controls static while cleaning and polishing. Effectively removes handling oils, finger soils, and nicotine stains. Prevents attraction of dust and dirt. Compatible with plastics, glass, and stainless steel. Foaming spray won't run or drip; ideal for optical surfaces. RoHS compliant.