Learn More
Electron Microscopy Sciences Foam Over Cotton Chemical Resistant Applicator Wooden Shaft, 13⁄16" Tip length
SDP
Catalog No. 50949170
Encompass_Preferred
Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time. Swabs meet all industry standards and requirements, including ASTM-D257 and Federal test method 4046, standard #101C.
OAL: 6"
Tip: 3⁄8"
Tip length: 13⁄16"
Catalog No. 50-949-170 Supplier Electron Microscopy Sciences Supplier No. 72316
Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time. Swabs meet all industry standards and requirements, including ASTM-D257 and Federal test method 4046, standard #101C.
OAL: 6"
Tip: 3⁄8"
Tip length: 13⁄16"