Learn More
Fisherbrand™ Antistatic Polyethylene Shoe Covers
Catalog No. 191300080 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Regular
X-Large
Sole Type:
Seamed Bottom
Seamless
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sole Type
19-130-0080 X-Large Seamless
19-098-933 X-Large Seamless
19-098-931 Regular Seamed Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-0080
This item is not returnable. View return policy
Offer exceptional durability
  • Inherently antistatic, fluid impervious, polyethylene-based film
  • Skid-resistant for added safety
  • Available with seamless bottom or with elastic seamed bottom for higher skid resistance
Order Info
Packed in double polybag.
Specifications
Size X-Large
Material Polyethylene
Closure Type Elastic Opening
Sole Type Seamless
Construction Polyethylene
Resistance Fluid
Product Type Shoe Covers