Learn More
MSC Delfin Industrial Accessory Kit, Antistatic, Wet/Dry, D80/50, 1 ea
Catalog No. 502867082
Encompass
This accessory kit is designed for the safe use of select vacuum cleaners, particularly when suctioning conductive dust and materials in both wet and dry applications. Its components are made of neoprene antistatic rubber and galvanized steel, ensuring correct earth grounding to reduce static.
  • Suitable for industrial and versatile applications
  • Allows for wet or dry cleaning
  • Ensures correct earth grounding to reduce static buildup
  • Includes brush strip, floor squeegee, lance, round brush, hose, and various connectors
  • Features an 80 mm vacuum diameter and 50 mm kit diameter
Catalog No. 50-286-7082 Supplier MSC Supplier No. 28841930
This accessory kit is designed for the safe use of select vacuum cleaners, particularly when suctioning conductive dust and materials in both wet and dry applications. Its components are made of neoprene antistatic rubber and galvanized steel, ensuring correct earth grounding to reduce static.
  • Suitable for industrial and versatile applications
  • Allows for wet or dry cleaning
  • Ensures correct earth grounding to reduce static buildup
  • Includes brush strip, floor squeegee, lance, round brush, hose, and various connectors
  • Features an 80 mm vacuum diameter and 50 mm kit diameter