Learn More
Sigma Aldrich Fine Chemicals Biosciences Weighing funnel, size XL, 10/pack
Catalog No. NC1809691
Encompass
Sit flat on balance - No rolling, no spilling!. Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer. Residual sample can be rinsed to guarantee 100% transfer. Suitable for safe handling of hazardous materials.
Catalog No. NC1809691 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5093611PAK
Sit flat on balance - No rolling, no spilling!. Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer. Residual sample can be rinsed to guarantee 100% transfer. Suitable for safe handling of hazardous materials.