Learn More
Medchemexpress LLC HY-W052512 100mg Medchemexpress, Antitrypanosomal agent 1 CAS:75144-12-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW052512100MG
Encompass_Preferred
Medchemexpress, HY-W052512 100mg Antitrypanosomal agent 1 CAS:75144-12-6 Antitrypanosomal agent 1 is a potent and selective trypanothione reductase (TR) inhibitor with an IC50 of 3.3 μM. Antitrypanosomal agent 1 inhibits glutathione reductase (GR) (IC50=64.8 μM) and T. brucei (EC50=1 μM). Antitrypanosomal agent 1 has anti-trypanosomal activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3489