Learn More
Medchemexpress LLC Antitumor agent-127 | 99.1% | 1670.92 | C86H107N15O18S | 10 MG
SDP
Catalog No. 5000288823
Encompass_Preferred
Antitumor agent-127 is a macrocyclic peptide ROR1 ligand supplied for research use. It shows nanomolar cellular binding and internalization in tumor cell lines, and is provided with supporting analytical documentation for characterization.
Catalog No. 50-002-88823 Supplier Medchemexpress LLC Supplier No. HY16109610MG
Antitumor agent-127 is a macrocyclic peptide ROR1 ligand supplied for research use. It shows nanomolar cellular binding and internalization in tumor cell lines, and is provided with supporting analytical documentation for characterization.