Learn More
Medchemexpress LLC Antitumor agent-159 10mg | 1611446-66-2 | 10 MG
SDP
Catalog No. 5000226178
Encompass_Preferred
Antitumor agent-159 is a mitochondria-targeting small-molecule research compound that downregulates cardiolipin, inhibits proliferation of MDA-MB-231 cancer cells, arrests the cell cycle at sub-G1, and induces apoptosis. It has demonstrated antitumor efficacy in MDA-MB-231 xenograft models. Molecular formula: C24H36ClF3N2O3; molecular weight: 493.00 g·mol⁻¹.
  • Targets mitochondria and reduces cardiolipin levels.
  • Inhibits proliferation of MDA-MB-231 cancer cells.
  • Arrests the cell cycle at sub-G1 and induces apoptosis.
  • Demonstrated in vivo efficacy in MDA-MB-231 xenograft models.
  • Intended for use in in vitro and in vivo antitumor research.
  • Reported purity: 99.0% (certificate of analysis).
Catalog No. 50-002-26178 Supplier Medchemexpress LLC Supplier No. HY11644710MG
Antitumor agent-159 is a mitochondria-targeting small-molecule research compound that downregulates cardiolipin, inhibits proliferation of MDA-MB-231 cancer cells, arrests the cell cycle at sub-G1, and induces apoptosis. It has demonstrated antitumor efficacy in MDA-MB-231 xenograft models. Molecular formula: C24H36ClF3N2O3; molecular weight: 493.00 g·mol⁻¹.
  • Targets mitochondria and reduces cardiolipin levels.
  • Inhibits proliferation of MDA-MB-231 cancer cells.
  • Arrests the cell cycle at sub-G1 and induces apoptosis.
  • Demonstrated in vivo efficacy in MDA-MB-231 xenograft models.
  • Intended for use in in vitro and in vivo antitumor research.
  • Reported purity: 99.0% (certificate of analysis).