Learn More
Electron Microscopy Sciences AMD-FH Microscope Workstation, 90 x 75 cm, Ferromagnetic SS Surface
SDP
Catalog No. 501930695
Encompass_Preferred
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Full Active table with self-levelling and platform guards.
Size: 90 x 75 cm
Surface: Ferromagnetic SS
Type: Active Air Self Leveling
Catalog No. 50-193-0695 Supplier Electron Microscopy Sciences Supplier No. 7513
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Full Active table with self-levelling and platform guards.
Size: 90 x 75 cm
Surface: Ferromagnetic SS
Type: Active Air Self Leveling