Learn More
Electron Microscopy Sciences AMD-MF Microscope Workstation, 120 x 75 cm, Stainless Steel Surface
SDP
Catalog No. 501930698
Encompass_Preferred
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Full width table guards.
Size: 120 x 75 cm
Surface: Stainless Steel
Type: Active Air Self Leveling
Catalog No. 50-193-0698 Supplier Electron Microscopy Sciences Supplier No. 7517
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Full width table guards.
Size: 120 x 75 cm
Surface: Stainless Steel
Type: Active Air Self Leveling