Learn More
Electron Microscopy Sciences AMD-MP Microscope Workstation, 120 x 75 cm, Surface
SDP
Catalog No. 501930701
Encompass_Preferred
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Simply pumped up via cycle valves. Full width table guards.
Size: 120 x 75 cm
Type: Active Air Self Leveling
Catalog No. 50-193-0701 Supplier Electron Microscopy Sciences Supplier No. 7720
These isolation tables support most of the larger microscopes and their support equipment. The air isolation platform effectively removes most removes most vibrations. The rigid frame has levelling feet which ensure the table is in excellent contact with the floor. The damped rolling diaphragm isolators have a low natural frequency of 2Hz which effectively isolates out vibrations above 6Hz, well below the frequencies at which typical building vibrations begin to occur. Simply pumped up via cycle valves. Full width table guards.
Size: 120 x 75 cm
Type: Active Air Self Leveling