Learn More
Electron Microscopy Sciences AMD-SM Microscope Workstation, 75 x 75 cm, Stainless Steel Surface
SDP
Catalog No. 501930705
Encompass_Preferred
Microscope workstations featuring a unique Sorbothane isolator design removing up to 98 percent of unwanted vibrations.
Size: 75 x 75 cm
Surface: Stainless Steel
Catalog No. 50-193-0705 Supplier Electron Microscopy Sciences Supplier No. 7506
Microscope workstations featuring a unique Sorbothane isolator design removing up to 98 percent of unwanted vibrations.
Size: 75 x 75 cm
Surface: Stainless Steel