Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antiviral agent 5 | 2698336-82-0 | 98.9% | 386.44 | 100 MG
SDP

Catalog No. 5000352182
Encompass_Preferred

Antiviral agent 5 is an intermediate used in antiviral agents targeting 3C and 3CL proteases, including SARS-CoV-2 Mpro. It is for research use only and not sold to patients.

  • Used in antiviral agents targeting 3C and 3CL proteases.
  • Targets SARS-CoV-2 Mpro.
  • For research use only.
  • Purity of 98.93%.
  • Molecular weight: 386.44.
  • Appears as an oil, colorless to light yellow.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-52182 Supplier Medchemexpress LLC Supplier No. HY139683100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antiviral agent 5 is an intermediate used in antiviral agents targeting 3C and 3CL proteases, including SARS-CoV-2 Mpro. It is for research use only and not sold to patients.

  • Used in antiviral agents targeting 3C and 3CL proteases.
  • Targets SARS-CoV-2 Mpro.
  • For research use only.
  • Purity of 98.93%.
  • Molecular weight: 386.44.
  • Appears as an oil, colorless to light yellow.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.